1-Chlor-5-iodpentan ≥97%

Lieferant: Thermo Scientific
Achtung

L04469.09
L04469.09EA 78.4 EUR
L04469.09
1-Chlor-5-iodpentan ≥97%
1-Chlor-5-iodpentan

Purity: 97% CAS No.: 60274-60-4 Molecular Formula: Cl(CH2)5I Formula Weight: 232.50 Flash Point: 110°(230°F) Refractive Index: 1.5305 Boiling Point: 75-77°/4mm Beilstein Reference: 1732342 MDL No.: MFCD00039416

Formel: C₅H₁₀ClI
Molecular Weight: 232,49 g/mol
Siedepunkt: 75…77 °C (4 mmHg)
Dichte: 1,663
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00039416
CAS-Nummer: 60274-60-4
EINECS: 262-137-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR