Triethanolamin (Trolamin, TEA) ≥98%

Lieferant: Thermo Fisher Scientific

L04486.22 L04486.36 L04486.0E
L04486.22EA 23.9 EUR
L04486.22 L04486.36 L04486.0E
Triethanolamin (Trolamin, TEA) ≥98%
Triethanolamin (Trolamin, TEA)

Purity: 98+% CAS No.: 102-71-6 Molecular Formula: C6H15NO3 Formula Weight: 149.19 Flash Point: 185°(365°F) Refractive Index: 1.4850 Odour: Ammonia-like Melting Point: 19-21° Boiling Point: 206-207°/15mm Beilstein Reference: 1699263 Merck Reference: 14,9665 MDL No.: MFCD00002855

Formel: C₆H₁₅NO₃
Siedepunkt: 206…207 °C (15 mmHg)
Schmelzpunkt: 19…21 °C
Dichte: 1,124
Flammpunkt: 185 °C (365 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002855
CAS-Nummer: 102-71-6
EINECS: 203-049-8
Merck Index: 14,9665
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=40c3d3d0c0162ac22208cadb8d5254a7cf0b340a&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR