2-Picolin-N-oxid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L04488.14
L04488.22
L04488.36
L04488.14EA
27.7
EUR
L04488.14
L04488.22
L04488.36
2-Picolin-N-oxid ≥98%
2-Picolin-N-oxid
Purity: 98%
CAS No.: 931-19-1
Molecular Formula: C6H7NO
Formula Weight: 109.13
Flash Point: 143°(289°F)
Melting Point: 46-50°
Boiling Point: 122°/8mm
Beilstein Reference: 106916
MDL No.: MFCD00006199
Formel:
C₆H₇NO Siedepunkt: 122 °C (8 mmHg) Schmelzpunkt: 46…50 °C Flammpunkt: 143 °C (289 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006199 CAS-Nummer: 931-19-1 EINECS: 213-230-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...