N-(3-Hydroxypropyl)phthalimid ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: N-(3-Hydroxypropyl)-1,2-benzoldicarbonsäureimid, N-(3-Hydroxypropyl)phthalsäureimid

L04507.09 L04507.18
L04507.09EA 29.7 EUR
L04507.09 L04507.18
N-(3-Hydroxypropyl)phthalimid ≥98%
N-(3-Hydroxypropyl)phthalimid

Purity: 98% CAS No.: 883-44-3 Molecular Formula: C11H11NO3 Formula Weight: 205.22 Melting Point: 75-80° Beilstein Reference: 158489

Formel: C₁₁H₁₁NO₃
Schmelzpunkt: 75…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023097
CAS-Nummer: 883-44-3
EINECS: 212-931-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR