(±)-1-Phenyl-1,2-ethandiol ≥97%

Lieferant: Thermo Fisher Scientific

L04551.06
L04551.06EA 42.5 EUR
L04551.06
(±)-1-Phenyl-1,2-ethandiol ≥97%
(±)-1-Phenyl-1,2-ethandiol

Purity: 97% CAS No.: 93-56-1 Molecular Formula: C8H10O2 Formula Weight: 138.17 Flash Point: 160°(320°F) Melting Point: 66-68° Boiling Point: 272-274° Beilstein Reference: 1306723 Merck Reference: 14,8861 MDL No.: MFCD00003546

Formel: C₆H₅CH(OH)CH₂OH
Siedepunkt: 272…274 °C
Schmelzpunkt: 66…68 °C
Flammpunkt: 160 °C (320 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003546
CAS-Nummer: 93-56-1
EINECS: 202-258-1
Merck Index: 14,8861

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR