(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
L-Cysteine ethyl ester hydrochloride, H-Cys-Oet.HCl, H-Cys-OEt·HCl
L04692.14
L04692.22
L04692.14EA
49
EUR
L04692.14
L04692.22
(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride ≥98%
(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride
Purity: 98+%
CAS No.: 868-59-7
Molecular Formula: C5H11NO2S.HCl
Formula Weight: 185.67
Melting Point: 123-125°
Beilstein Reference: 3562600
Optical Rotation: -12° (c=1 in 1N HCl)
MDL No.: MFCD00012631
Formel:
C₅H₁₁NO₂S·HCl Schmelzpunkt: 123…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012631 CAS-Nummer: 868-59-7 EINECS: 212-779-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...