Bisacrylamid ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N,N'-Methylendiacrylamid, BIS
L04694.22
L04694.36
L04694.22EA
41.7
EUR
L04694.22
L04694.36
Bisacrylamid ≥97%
Bisacrylamid
Purity: 97%
CAS No.: 110-26-9
Molecular Formula: C7H10N2O2
Formula Weight: 154.17
Storage Temperature: 2°C to 8°C
Melting Point:
Formel:
(H₂C=CHCONH)₂CH₂ Siedepunkt: 125 °C (3,33 hPa) Schmelzpunkt: >300 °C Dichte: 1,235 Flammpunkt: 138 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00008625 CAS-Nummer: 110-26-9 EINECS: 203-750-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...