1-(3-Aminopropyl)pyrrolidin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L04739.06
L04739.14
L04739.06EA
67.5
EUR
L04739.06
L04739.14
1-(3-Aminopropyl)pyrrolidin 97%
Purity: 97%
CAS No.: 23159-07-1
Molecular Formula: C7H16N2
Formula Weight: 128.22
Refractive Index: 1.4710
Boiling Point: 191-193°
UN No.: 2735
MDL No.: MFCD00014100
Formel:
C₇H₁₆N₂ Siedepunkt: 191…193 °C Dichte: 1,014 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014100 CAS-Nummer: 23159-07-1 EINECS: 245-465-2 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...