3-Chlor-α-methylbenzylalkohol ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: 3-Chlorophenyl methyl carbinol, 1-(3-Chlorophenyl)ethanol, m-Chloro-α-methylbenzyl alcohol

L04783.06 L04783.14
L04783.06EA 43 EUR
L04783.06 L04783.14
3-Chlor-α-methylbenzylalkohol ≥97%
3-Chlor-α-methylbenzylalkohol

Purity: 97% CAS No.: 6939-95-3 Molecular Formula: C8H9ClO Formula Weight: 156.61 Flash Point: 110°(230°F) Refractive Index: 1.5445 Boiling Point: 83-85°/1.5mm Beilstein Reference: 2042021 MDL No.: MFCD00021864

Formel: C₈H₉ClO
Molecular Weight: 156.61 g/mol
Siedepunkt: 83…85 °C (1.5 mmHg)
Dichte: 1.173
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00021864
CAS-Nummer: 6939-95-3
EINECS: 230-079-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR