3-Chlor-α-methylbenzylalkohol ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme:
3-Chlorophenyl methyl carbinol, 1-(3-Chlorophenyl)ethanol, m-Chloro-α-methylbenzyl alcohol
L04783.06
L04783.14
L04783.06EA
43
EUR
L04783.06
L04783.14
3-Chlor-α-methylbenzylalkohol ≥97%
3-Chlor-α-methylbenzylalkohol
Purity: 97%
CAS No.: 6939-95-3
Molecular Formula: C8H9ClO
Formula Weight: 156.61
Flash Point:
Formel:
C₈H₉ClO Molecular Weight: 156.61 g/mol Siedepunkt: 83…85 °C (1.5 mmHg) Dichte: 1.173 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00021864 CAS-Nummer: 6939-95-3 EINECS: 230-079-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...