δ-Valerolacton ≥98% may contain polymer

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Pentansäure-δ-lacton, 5-Hydroxyvaleriansäurelacton, Tetrahydro-2H-pyran-2-on, Pentan-5-olid

L04794.14 L04794.22 L04794.36
L04794.14EA 54.3 EUR
L04794.14 L04794.22 L04794.36
δ-Valerolacton ≥98% may contain polymer
δ-Valerolacton

Purity: 98% CAS No.: 542-28-9 Molecular Formula: C5H8O2 Formula Weight: 100.12 Storage Temperature: 2°C to 8°C Flash Point: 112°(233°F) Refractive Index: 1.4580 Melting Point: -13 to -12° Boiling Point: 58-60°/0.5mm Beilstein Reference: 106436 MDL No.: MFCD00006645

Formel: C₅H₈O₂
Molecular Weight: 100.12 g/mol
Siedepunkt: 58…60 °C (0.5 mmHg)
Schmelzpunkt: –13 °C…–12 °C
Dichte: 1.056
Flammpunkt: 112 °C (233 °F)
MDL: MFCD00006645
CAS-Nummer: 542-28-9
EINECS: 208-807-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR