3-(3-Indolyl)propionsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L04877.14 L04877.06
L04877.14EA 84.6 EUR
L04877.14 L04877.06
3-(3-Indolyl)propionsäure ≥98%
3-(3-Indolyl)propionsäure

Purity: 98% CAS No.: 830-96-6 Molecular Formula: C11H11NO2 Formula Weight: 189.21 Melting Point: 132-135° Beilstein Reference: 147733

Formel: C₁₁H₁₁NO₂
Schmelzpunkt: 132…135 °C
Dichte: 1,356 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005660
CAS-Nummer: 830-96-6
EINECS: 212-600-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR