3-Acetoxyindol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Indolyl Acetate, Indoxyl acetate, 3-Indoxyl acetate, 3-Indolyl acetate
L04932.03
L04932.03EA
48.9
EUR
L04932.03
3-Acetoxyindol ≥97%
3-Acetoxyindol
Purity: 97%
CAS No.: 608-08-2
Molecular Formula: C10H9NO2
Formula Weight: 175.19
Melting Point: 128-130°
Beilstein Reference: 143086
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol Schmelzpunkt: 128…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014561 CAS-Nummer: 608-08-2 EINECS: 210-154-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...