Cell culture media, RPMI 1640
Lieferant: Biowest
L0490-500
L0492-500
L0495-500
L0496-500
L0498-500
L0500-100
L0500-500
L0501-100
L0501-500
P0860-N1L
P0860-N10L
P0870-N1L
P0870-N10L
P0871-N1L
P0880-N10L
P0883-N1L
P0870-N50L
P0876-N1L
P0876-N10L
P0876-N50L
P0880-N50L
P0883-N10L
P0860-N5L
P0860-N50L
P0880-N1L
P0871-N10L
L0498-100
L0503-500
L0505-500
L0501-1000
L0490-500EA
12.4
EUR
L0490-500
L0492-500
L0495-500
L0496-500
L0498-500
L0500-100
L0500-500
L0501-100
L0501-500
P0860-N1L
P0860-N10L
P0870-N1L
P0870-N10L
P0871-N1L
P0880-N10L
P0883-N1L
P0870-N50L
P0876-N1L
P0876-N10L
P0876-N50L
P0880-N50L
P0883-N10L
P0860-N5L
P0860-N50L
P0880-N1L
P0871-N10L
L0498-100
L0503-500
L0505-500
L0501-1000
Cell culture media, RPMI 1640
Zellkulturmedien
RPMI are general purpose enriched media with extensive applications for a broad spectrum of mammalian and hybridoma cells, including human myeloma, mouse hybridoma, human leukocytes, and B and T lymphocytes. It was originally formulated for suspension cultures and monolayer culture of human leukaemia cells.