Cinnamaldoxime Gemisch aus EZ-Isomeren 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Cinnamaldehyde oxime

L05026.06 L05026.14
L05026.06EA 37.5 EUR
L05026.06 L05026.14
Cinnamaldoxime Gemisch aus EZ-Isomeren 98%
Cinnamaldoxime

Purity: 98% CAS No.: 13372-81-1 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 117-122° Beilstein Reference: 3117555 MDL No.: MFCD00019969

Formel: C₉H₉NO
Schmelzpunkt: 117…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00019969
CAS-Nummer: 13372-81-1
EINECS: 236-449-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR