1,1-Dimethylharnstoff ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L05065.09
L05065.09EA
45.4
EUR
L05065.09
1,1-Dimethylharnstoff ≥98%
1,1-Dimethylharnstoff
Purity: 98%
CAS No.: 598-94-7
Molecular Formula: (CH3)2NCONH2
Formula Weight: 88.11
Melting Point: 178-183°
Beilstein Reference: 1740666
Formel:
C₃H₈N₂O Schmelzpunkt: 178…183 °C Dichte: 1,255 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007959 CAS-Nummer: 598-94-7 EINECS: 209-957-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...