7-Bromheptannitril ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-Brom-6-cyanhexan, 7-Bromheptansäurenitril, 7-Bromönanthsäurenitril

L05068.06
L05068.06EA 52.5 EUR
L05068.06
7-Bromheptannitril ≥98%
7-Bromheptannitril

Purity: 98% CAS No.: 20965-27-9 Molecular Formula: Br(CH2)6CN Formula Weight: 190.09 Flash Point: 110°(230°F) Refractive Index: 1.4750 Boiling Point: 114-115°/4mm Beilstein Reference: 1748070 UN No.: 3276 MDL No.: MFCD00001982

Formel: BrH₂C(CH₂)₄CH₂CN
Molecular Weight: 190,08 g/mol
Siedepunkt: 114…115 °C (4 mmHg)
Dichte: 1,266
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001982
CAS-Nummer: 20965-27-9
EINECS: 244-128-7
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR