1-(2-Naphthyl)ethanol 97%
Lieferant: Thermo Scientific
Synonyme:
alpha-Methyl-2-naphthalenemethanol, 2-(1-Hydroxyethyl)naphthalene
L05108.09
L05108.09EA
92.5
EUR
L05108.09
1-(2-Naphthyl)ethanol 97%
1-(2-Naphthyl)ethanol
Purity: 97%
CAS No.: 7228-47-9
Molecular Formula: C12H12O
Formula Weight: 172.23
Melting Point: 72-75°
Boiling Point: 170°/13mm
Beilstein Reference: 1907449
MDL No.: MFCD00004111
Formel:
C₁₂H₁₂O Molecular Weight: 172,23 g/mol Siedepunkt: 170 °C (13 mmHg) Schmelzpunkt: 72…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004111 CAS-Nummer: 7228-47-9 EINECS: 230-630-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...