11-Heneicosanol 97%

Lieferant: Thermo Fisher Scientific

L05187.03 L05187.06
L05187.03EA 77.5 EUR
L05187.03 L05187.06
11-Heneicosanol 97%
11-Heneicosanol

Purity: 97% CAS No.: 3381-26-8 Molecular Formula: C21H44O Formula Weight: 312.58 Odour: Odourless Melting Point: 71-73° Beilstein Reference: 1765841 MDL No.: MFCD00026547

Formel: C₂₁H₄₄O
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00026547
CAS-Nummer: 3381-26-8
EINECS: 222-184-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR