4-Cyan-4-phenylcyclohexanon 97%

Lieferant: Thermo Fisher Scientific
Achtung

L05281.06
L05281.06EA 76.6 EUR
L05281.06
4-Cyan-4-phenylcyclohexanon 97%
4-Cyano-4-phenylcyclohexanone

Purity: 98% CAS No.: 25115-74-6 Molecular Formula: C13H13NO Formula Weight: 199.24 Melting Point: 112-118° Beilstein Reference: 2645593

Formel: C₁₃H₁₃NO
Schmelzpunkt: 112…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00044816
CAS-Nummer: 25115-74-6
EINECS: 246-631-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR