Methyl-3-phenylpropionat ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Methyldihydrocinnamat, 3-Phenylpropionsäuremethylester, Hydrozimtsäuremethylester

L05310.18 L05310.30
L05310.18EA 99.9 EUR
L05310.18 L05310.30
Methyl-3-phenylpropionat ≥98%
Methyl-3-phenylpropionat

Purity: 98% CAS No.: 103-25-3 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 100°(212°F) Refractive Index: 1.5020 Boiling Point: 236-239° Beilstein Reference: 1909591 MDL No.: MFCD00017209

Formel: C₁₀H₁₂O₂
Molecular Weight: 164.2 g/mol
Siedepunkt: 236…239 °C
Dichte: 1.045
Flammpunkt: 100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00017209
CAS-Nummer: 103-25-3
EINECS: 203-092-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR