4-Ethoxybenzhydrazid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Ethoxybenzoic hydrazide

L05433.06
L05433.06EA 63.6 EUR
L05433.06
4-Ethoxybenzhydrazid ≥98%
4-Ethoxybenzhydrazid

Purity: 98+% CAS No.: 58586-81-5 Molecular Formula: C9H12N2O2 Formula Weight: 180.21 Melting Point: 124-127° Beilstein Reference: 2642900

Formel: C₉H₁₂N₂O₂
Molecular Weight: 180.21 g/mol
Schmelzpunkt: 124…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017074
CAS-Nummer: 58586-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR