2-(Methylthio)ethanol ≥99%
Lieferant: Thermo Fisher Scientific
L05442.06
L05442.14
L05442.22
L05442.06EA
24.6
EUR
L05442.06
L05442.14
L05442.22
2-(Methylthio)ethanol ≥99%
Purity: 99%
CAS No.: 5271-38-5
Molecular Formula: C3H8OS
Formula Weight: 92.16
Flash Point: 70°(158°F)
Refractive Index: 1.4944
Odour: Stench
Boiling Point: 169-171°
Beilstein Reference: 1731081
UN No.: 3334
MDL No.: MFCD00002908
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...