Tetradecyltrichlorsilan ≥97%
Lieferant: Thermo Scientific
Synonyme:
Trichloro(tetradecyl)silane
L05495.03
L05495.03EA
0
EUR
L05495.03
Tetradecyltrichlorsilan ≥97%
Tetradecyltrichlorsilan
Purity: 97%
CAS No.: 18402-22-7
Molecular Formula: CH3(CH2)13SiCl3
Formula Weight: 331.83
Refractive Index: 1.4565
Boiling Point: 155-157°/3mm
UN No.: 2987
MDL No.: MFCD00051440
Formel:
C₁₄H₂₉Cl₃Si Molecular Weight: 331,83 g/mol Siedepunkt: 131…132 °C (0,8 mmHg) Dichte: 1 g/cm³ (20 °C) |
MDL:
MFCD00051440 CAS-Nummer: 18402-22-7 UN: 2987 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...