2,5-Diaminopyridin Dihydrochlorid 97%

Lieferant: Thermo Scientific
Achtung

L05599.06
L05599.06EA 47.9 EUR
L05599.06
2,5-Diaminopyridin Dihydrochlorid 97%
2,5-Diaminopyridin Dihydrochlorid

Purity: 97% CAS No.: 26878-35-3 Molecular Formula: C5H7N3.2HCl Formula Weight: 182.05 Melting Point: ca 264° dec. Beilstein Reference: 3912365

Formel: C₅H₉Cl₂N₃
Molecular Weight: 182,05 g/mol
Schmelzpunkt: ∼ 264 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00040818
CAS-Nummer: 26878-35-3
EINECS: 248-083-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR