n-Nonadecan ≥99%

Lieferant: Thermo Fisher Scientific

L05701.14 L05701.22
L05701.14EA 36.4 EUR
L05701.14 L05701.22
n-Nonadecan ≥99%
n-Nonadecan

Purity: 99% CAS No.: 629-92-5 Molecular Formula: CH3(CH2)17CH3 Formula Weight: 268.53 Flash Point: 168°(334°F) Refractive Index: 1.4410 Melting Point: 32-34° Boiling Point: 330-331° Beilstein Reference: 1742892 MDL No.: MFCD00009012

Formel: H₃C(CH₂)₁₇CH₃
Siedepunkt: 330…331 °C
Schmelzpunkt: 32…34 °C
Dichte: 0,786
Flammpunkt: 168 °C (334 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009012
CAS-Nummer: 629-92-5
EINECS: 211-116-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR