5-Chlorothiazol-2-amine hydrochloride 97%

Lieferant: Thermo Fisher Scientific
Gefahr

L05707.06 L05707.03 L05707.14
L05707.06EA 93.4 EUR
L05707.06 L05707.03 L05707.14
5-Chlorothiazol-2-amine hydrochloride 97%
5-Chlorothiazol-2-amine hydrochloride

Purity: 97% CAS No.: 55506-37-1 Molecular Formula: C3H3ClN2S.HCl Formula Weight: 171.05 Melting Point: 168-172° MDL No.: MFCD00039006

Formel: C3H4Cl2N2S
Schmelzpunkt: 168…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039006
CAS-Nummer: 55506-37-1
EINECS: 259-680-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR