3-Ethoxybenzoesäure ≥98%

Lieferant: Thermo Scientific
Achtung

L05721.09
L05721.09EA 96.9 EUR
L05721.09
3-Ethoxybenzoesäure ≥98%
3-Ethoxybenzoic acid

Purity: 98+% CAS No.: 621-51-2 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 135-137° Beilstein Reference: 509159

Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Schmelzpunkt: 135…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016517
CAS-Nummer: 621-51-2
EINECS: 210-690-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR