5-Nonanol ≥98%

Lieferant: Thermo Fisher Scientific

L05812.14
L05812.14EA 47.1 EUR
L05812.14
5-Nonanol ≥98%
5-Nonanol

Purity: 98% CAS No.: 623-93-8 Molecular Formula: C9H20O Formula Weight: 144.26 Flash Point: 96°(204°F) Refractive Index: 1.4290 Melting Point: 5° Boiling Point: 194-196° Beilstein Reference: 1733552 MDL No.: MFCD00021944

Formel: C₉H₂₀O
Molecular Weight: 144,26 g/mol
Siedepunkt: 194…196 °C
Schmelzpunkt: 5 °C
Dichte: 0,826
Flammpunkt: 96 °C (204 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00021944
CAS-Nummer: 623-93-8
EINECS: 210-820-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR