trans-2-Decenal 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (E)-2-Decenal

L05901.06 L05901.14
L05901.06EA 55.7 EUR
L05901.06 L05901.14
trans-2-Decenal 95%
trans-2-Decenal

Purity: 95% CAS No.: 3913-81-3 Molecular Formula: C10H18O Formula Weight: 154.25 Storage Temperature: 2°C to 8°C Flash Point: 93°(199°F) Refractive Index: 1.4565 Boiling Point: 117°/17mm Beilstein Reference: 1704445 MDL No.: MFCD00014679

Formel: C₁₀H₁₈O
Siedepunkt: 117 °C (17 mmHg)
Dichte: 0,840
Flammpunkt: 93 °C (199 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00014679
CAS-Nummer: 3913-81-3
EINECS: 223-474-2
UN: 3082
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR