Methyl-3,4,5-trihydroxybenzoat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,4,5-Trihydroxybenzoesäuremethylester, Methylgallat, Gallussäuremethylester
L05926.18
L05926.30
L05926.0250
L05926.18EA
49.3
EUR
L05926.18
L05926.30
ABCRL05926.0250
Methyl-3,4,5-trihydroxybenzoat ≥98%
Methyl-3,4,5-trihydroxybenzoat
Purity: 98%
CAS No.: 99-24-1
Molecular Formula: C8H8O5
Formula Weight: 184.15
Melting Point: 198-203°
Beilstein Reference: 2113180
Merck Reference: 14,4345
MDL No.: MFCD00002194
Formel:
C₈H₈O₅ Molecular Weight: 184.15 g/mol Siedepunkt: 450 Schmelzpunkt: 198…203 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002194 CAS-Nummer: 99-24-1 EINECS: 202-741-7 Merck Index: 13,04366 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...