DL-Tryptophan ≥99%

Lieferant: Thermo Fisher Scientific

L05936.06 L05936.14 L05936.22
L05936.06EA 25.6 EUR
L05936.06 L05936.14 L05936.22
DL-Tryptophan ≥99%
DL-Tryptophan

Purity: 99% CAS No.: 54-12-6 Molecular Formula: C11H12N2O2 Formula Weight: 204.23 Melting Point: ca 280° dec. Beilstein Reference: 86196

Formel: C₁₁H₁₂N₂O₂
Dichte: 1,349 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064339
CAS-Nummer: 54-12-6
EINECS: 200-194-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR