Methyl-5-amino-2-furoat 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 5-aminofuran-2-carboxylate

L05958.03 L05958.06
L05958.03EA 96 EUR
L05958.03 L05958.06
Methyl-5-amino-2-furoat 98%
Methyl-5-amino-2-furoat

Purity: 98% CAS No.: 22600-30-2 Molecular Formula: C6H7NO3 Formula Weight: 141.13 Melting Point: 131-135° Beilstein Reference: 1365799

Formel: C₆H₇NO₃
Schmelzpunkt: 131…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051661
CAS-Nummer: 22600-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR