2-Chlor-6-methoxypyridin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L05997.14
L05997.06
L05997.14EA
99.4
EUR
L05997.14
L05997.06
2-Chlor-6-methoxypyridin ≥98%
2-Chloro-6-methoxypyridine
Purity: 98%
CAS No.: 17228-64-7
Molecular Formula: C6H6ClNO
Formula Weight: 143.57
Flash Point: 76°(168°F)
Refractive Index: 1.5280
Boiling Point: 185-186°
Beilstein Reference: 1364500
MDL No.: MFCD00006265
Formel:
C₆H₆ClNO Siedepunkt: 185…186 °C Dichte: 1,207 Flammpunkt: 76 °C (168 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006265 CAS-Nummer: 17228-64-7 EINECS: 241-264-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...