4'-Chlor-3'-methylacetophenon ≥75% remainder 3'-chloro-4'-methylacetophenone, tech.

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(4-Chloro-3-methylphenyl)ethanone

L06011.06 L06011.14
L06011.06EA 50.9 EUR
L06011.06 L06011.14
4'-Chlor-3'-methylacetophenon ≥75% remainder 3'-chloro-4'-methylacetophenone, tech.
4'-Chlor-3'-methylacetophenon

Purity: 75% CAS No.: 37074-39-8 Molecular Formula: C9H9ClO Formula Weight: 168.62 Flash Point: 110°(230°F) Boiling Point: 104-105°/0.4mm MDL No.: MFCD00045172

Formel: C₉H₉ClO
Molecular Weight: 168.62 g/mol
Siedepunkt: 104…105 °C (0.4 mmHg)
Dichte: 1.15
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00045172
CAS-Nummer: 37074-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR