1,2,6-Hexantriol 96%

Lieferant: Thermo Fisher Scientific

Synonyme: 1,2,6-Trihydroxyhexane

L06032.14
L06032.14EA 46.2 EUR
L06032.14
1,2,6-Hexantriol 96%
1,2,6-Hexantriol

Purity: 96% CAS No.: 106-69-4 Molecular Formula: C6H14O3 Formula Weight: 134.18 Flash Point: 191°(375°F) Refractive Index: 1.4760 Melting Point: -20° Boiling Point: 166-168°/2mm Beilstein Reference: 1304479 MDL No.: MFCD00002976

Formel: C₆H₁₄O₃
Molecular Weight: 134.18 g/mol
Siedepunkt: 166…168 °C (2 mmHg)
Schmelzpunkt: –20 °C
Dichte: 1.106
Flammpunkt: 191 °C (375 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002976
CAS-Nummer: 106-69-4
EINECS: 203-424-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR