2-(4-Methoxyphenyl)ethanol ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-(4-Methoxyphenyl)ethylalkohol, 4-Methoxyphenethylalkohol

L06158.09 L06158.18
L06158.09EA 42.2 EUR
L06158.09 L06158.18
2-(4-Methoxyphenyl)ethanol ≥98%
2-(4-Methoxyphenyl)ethanol

Purity: 98% CAS No.: 702-23-8 Molecular Formula: C9H12O2 Formula Weight: 152.19 Flash Point: 110°(230°F) Refractive Index: 1.5370 Melting Point: 26-30° Boiling Point: 334-336° Beilstein Reference: 2043563 MDL No.: MFCD00002900

Formel: C₉H₁₂O₂
Siedepunkt: 334…336 °C
Schmelzpunkt: 26…30 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002900
CAS-Nummer: 702-23-8
EINECS: 211-866-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR