4,5-Dimethoxyanthranilsäure 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Amino-4,5-dimethoxybenzoesäure, 6-Aminoveratrumsäure, 6-Aminoveratrinsäure
L06205.14
L06205.14EA
71.5
EUR
L06205.14
4,5-Dimethoxyanthranilsäure 98%
4,5-Dimethoxyanthranilsäure
Purity: 98%
CAS No.: 5653-40-7
Molecular Formula: C9H11NO4
Formula Weight: 197.19
Melting Point: ca 155-158° dec.
Beilstein Reference: 782814
Formel:
C₉H₁₁NO₄ Molecular Weight: 197.19 g/mol Siedepunkt: 359 Schmelzpunkt: ∼155…158 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011671 CAS-Nummer: 5653-40-7 EINECS: 227-095-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...