(Dimethylamino)acetaldehyddiethylacetal 95%

Lieferant: Thermo Fisher Scientific
Achtung

L06272.14
L06272.14EA 107 EUR
L06272.14
(Dimethylamino)acetaldehyddiethylacetal 95%
(Dimethylamino)acetaldehyddiethylacetal

Purity: 95% CAS No.: 3616-56-6 Molecular Formula: (CH3)2NCH2CH(OCH2CH3)2 Formula Weight: 161.25 Flash Point: 45°(113°F) Refractive Index: 1.4115 Odour: Amine-like Boiling Point: 164-166° Beilstein Reference: 1739149 UN No.: 1993 MDL No.: MFCD00009232

Formel: C₈H₁₉NO₂
Siedepunkt: 164…166 °C
Dichte: 0,876
Flammpunkt: 45 °C (113 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009232
CAS-Nummer: 3616-56-6
EINECS: 222-800-0
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR