L(+)-Cystein Hydrochlorid, wasserfrei 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
H-Cys-OH.HCl, (R)-(+)-Cystein Hydrochlorid, (R)-(+)-2-Amino-3-mercaptopropansäure Hydrochlorid
L06328.14
L06328.14EA
48.3
EUR
L06328.14
L(+)-Cystein Hydrochlorid, wasserfrei 98%
L(+)-Cystein Hydrochlorid
Purity: 98%
CAS No.: 52-89-1
Molecular Formula: C3H7NO2S.HCl
Formula Weight: 157.62
Odour: Irritating
pH: 0.8-1.2
Melting Point: ca 180° dec.
Beilstein Reference: 3560277
Merck Reference: 14,2781
Optical Rotation: +6.4° (c=5 in 5N HCl)
MDL No.: MFCD00064553
Formel:
HSCH₂CH(NH₂)COOH·HCl Molecular Weight: 157,62 g/mol Schmelzpunkt: ∼ 180 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064553 CAS-Nummer: 52-89-1 EINECS: 200-157-7 Merck Index: 14,2781 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...