5-Chlor-2-(methylamino)benzophenon 99%

Lieferant: Thermo Scientific

Synonyme: (5-Chloro-2-(methylamino)phenyl)(phenyl)methanone

L06338.09
L06338.09EA 32.1 EUR
L06338.09
5-Chlor-2-(methylamino)benzophenon 99%
5-Chlor-2-(methylamino)benzophenon

Purity: 99% CAS No.: 1022-13-5 Molecular Formula: C14H12ClNO Formula Weight: 245.71 Melting Point: 93-98° Beilstein Reference: 882187

Formel: C₁₄H₁₂ClNO
Molecular Weight: 245,71 g/mol
Schmelzpunkt: 93…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008284
CAS-Nummer: 1022-13-5
EINECS: 213-822-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR