4-(Aminomethyl)pyridin ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Picolamin, 4-Picolylamin, 4-Pyridylmethylamin, (4-Pyridyl)methylamin
L06345.14
L06345.22
L06345.14EA
53.5
EUR
L06345.14
L06345.22
4-(Aminomethyl)pyridin ≥97%
4-(Aminomethyl)pyridine
Purity: 97+%
CAS No.: 3731-53-1
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 108°(226°F)
Refractive Index: 1.5515
Odour: Amine-like
Melting Point: -8°
Boiling Point: 230°
Beilstein Reference: 108057
UN No.: 2735
MDL No.: MFCD00006449
Formel:
C₆H₈N₂ Siedepunkt: 230 °C Schmelzpunkt: –8 °C Dichte: 1,069 Flammpunkt: 108 °C (226 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006449 CAS-Nummer: 3731-53-1 EINECS: 223-092-6 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...