4-Chlorbutyrophenon ≥95%
Lieferant: Thermo Scientific
Synonyme:
γ-Chlorbutyrophenon, ω-Chlorbutyrophenon
L06383.14
L06383.14EA
158
EUR
L06383.14
4-Chlorbutyrophenon ≥95%
4-Chlorbutyrophenon
Purity: 95%
CAS No.: 939-52-6
Molecular Formula: C10H11ClO
Formula Weight: 182.65
Flash Point:
Formel:
(C₆H₅)C(O)(CH₂)₃Cl Molecular Weight: 182,65 g/mol Siedepunkt: 138…140 °C (5 mmHg) Schmelzpunkt: 19…20 °C Dichte: 1,137 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001005 CAS-Nummer: 939-52-6 EINECS: 213-362-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...