1,7-Heptandiol ≥98%
Lieferant: Thermo Scientific
Synonyme:
Heptamethylenglycol, 1,7-Dihydroxyheptan, Heptamethylenglykol
L06384.06
L06384.06EA
55.2
EUR
L06384.06
1,7-Heptandiol ≥98%
1,7-Heptandiol
Purity: 98%
CAS No.: 629-30-1
Molecular Formula: HO(CH2)7OH
Formula Weight: 132.20
Flash Point: 127°(260°F)
Refractive Index: 1.4550
Melting Point: 17-18°
Boiling Point: 258-260°
Beilstein Reference: 1633482
MDL No.: MFCD00002987
Formel:
C₇H₁₆O₂ Molecular Weight: 132,2 g/mol Siedepunkt: 258…260 °C Schmelzpunkt: 17…18 °C Dichte: 0,953 Flammpunkt: 127 °C (260 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002987 CAS-Nummer: 629-30-1 EINECS: 211-085-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...