1-Phenyl-4,5-dihydro-1H-pyrazol-3-ylamine ≥98%

Lieferant: Thermo Scientific
Achtung

L06476.09
L06476.09EA 52.6 EUR
L06476.09
1-Phenyl-4,5-dihydro-1H-pyrazol-3-ylamine ≥98%
1-Phenyl-4,5-dihydro-1H-pyrazol-3-ylamine

Purity: 98+% CAS No.: 3314-35-0 Molecular Formula: C9H11N3 Formula Weight: 161.21 Melting Point: 167-170° Beilstein Reference: 139162 MDL No.: MFCD00051730

Formel: C₉H₁₁N₃
Molecular Weight: 161,21 g/mol
Schmelzpunkt: 167…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051730
CAS-Nummer: 3314-35-0
EINECS: 222-005-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR