5-Amino-1H-indazol 98%

Lieferant: Thermo Fisher Scientific
Gefahr

L06705.06 L06705.14
L06705.06EA 47 EUR
L06705.06 L06705.14
5-Amino-1H-indazol 98%
5-Amino-1H-indazole

Purity: 98% CAS No.: 19335-11-6 Molecular Formula: C7H7N3 Formula Weight: 133.15 Odour: Odourless Melting Point: 172-178° Beilstein Reference: 3259 UN No.: 2811 MDL No.: MFCD00037975

Formel: C₇H₇N₃
Siedepunkt: 345 °C (1013 hPa)
Schmelzpunkt: 172…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00037975
CAS-Nummer: 19335-11-6
EINECS: 242-971-5
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR