4-Fluorbenzamid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Fluorbenzoesäureamid

L06831.14 L06831.06 L06831.22
L06831.14EA 93.4 EUR
L06831.14 L06831.06 L06831.22
4-Fluorbenzamid ≥98%
4-Fluorbenzamid

Purity: 98% CAS No.: 824-75-9 Molecular Formula: C7H6FNO Formula Weight: 139.13 Melting Point: 154-157° Beilstein Reference: 1859744

Formel: C₇H₆FNO
Molecular Weight: 139.13 g/mol
Siedepunkt: 287
Schmelzpunkt: 154…157 °C
Dichte: 1.43 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007992
CAS-Nummer: 824-75-9
EINECS: 212-535-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR