4-Fluorbenzamid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Fluorbenzoesäureamid
L06831.14
L06831.06
L06831.22
L06831.14EA
93.4
EUR
L06831.14
L06831.06
L06831.22
4-Fluorbenzamid ≥98%
4-Fluorbenzamid
Purity: 98%
CAS No.: 824-75-9
Molecular Formula: C7H6FNO
Formula Weight: 139.13
Melting Point: 154-157°
Beilstein Reference: 1859744
Formel:
C₇H₆FNO Molecular Weight: 139.13 g/mol Siedepunkt: 287 Schmelzpunkt: 154…157 °C Dichte: 1.43 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007992 CAS-Nummer: 824-75-9 EINECS: 212-535-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...