2-(Methylthio)benzonitril ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

L06870.06 L06870.03
L06870.06EA 194 EUR
L06870.06 L06870.03
2-(Methylthio)benzonitril ≥98%
2-(Methylthio)benzonitril

Purity: 98% CAS No.: 6609-54-7 Molecular Formula: C8H7NS Formula Weight: 149.22 Melting Point: 33-38° Boiling Point: 116-117°/5mm UN No.: 3439 MDL No.: MFCD00015557

Formel: C₈H₇NS
Molecular Weight: 149.22 g/mol
Siedepunkt: 116…117 °C (5 mmHg)
Schmelzpunkt: 33…38 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015557
CAS-Nummer: 6609-54-7
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR