1-(2-Aminophenyl)pyrrol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

L06887.03 L06887.06
L06887.03EA 26.6 EUR
L06887.03 L06887.06
1-(2-Aminophenyl)pyrrol ≥98%
1-(2-Aminophenyl)pyrrole

Purity: 98+% CAS No.: 6025-60-1 Molecular Formula: C10H10N2 Formula Weight: 158.20 Melting Point: 93-94° Beilstein Reference: 1307631

Formel: C₁₀H₁₀N₂
Schmelzpunkt: 93…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005344
CAS-Nummer: 6025-60-1
EINECS: 227-884-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR